missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Diphenylphosphinite, TRC

Product Code. 30369490
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30369490 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369490 Supplier Toronto Research Chemicals Supplier No. TRCE937100500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Diphenylphosphinite
CAS 719-80-2
Molecular Formula C14H15OP
Quantity 500 mg
InChI Formula InChI=1S/C14H15OP/c1-2-15-16(13-9-5-3-6-10-13)14-11-7-4-8-12-14/h3-12H,2H2,1H3
SMILES CCOP(c1ccccc1)c2ccccc2
IUPAC Name ethoxy(diphenyl)phosphane
Molecular Weight (g/mol) 230.24
Formula Weight 230.0861
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.