missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (E)-3-(1-pyrrolidinyl)crotonate, 97%

Product Code. 11330054
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11330054 25 g 25g
11320054 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11330054 Supplier Thermo Scientific Alfa Aesar Supplier No. L00461.14
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 54716-02-8 | C10H17NO2 | 183.251 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54716-02-8
Molecular Formula C10H17NO2
Molecular Weight (g/mol) 183.251
MDL Number MFCD00014097
InChI Key MSOQKPXSIHLODG-CMDGGOBGSA-N
Synonym e-ethyl 3-pyrrolidin-1-yl but-2-enoate, ethyl e-3-1-pyrrolidinyl crotonate, 2e-3-1-pyrrolidinyl-2-butenoic acid ethyl ester, ethyl e-3-1-pyrrolidino crotonate, ethyl e-3-pyrrolidin-1-ylbut-2-enoate, e-3-pyrrolidin-1-yl-but-2-enoic acid ethyl ester, 2-butenoic acid, 3-1-pyrrolidinyl-, ethyl ester, e, ethyl 2e-3-pyrrolidin-1-yl but-2-enoate, ethyl 3-1-pyrrolidinyl crotonate
PubChem CID 736203
IUPAC Name ethyl (E)-3-pyrrolidin-1-ylbut-2-enoate
SMILES CCOC(=O)C=C(C)N1CCCC1

Specifications

Boiling Point 125°C to 126°C (0.8 mmHg)
Refractive Index 1.541
Quantity 25 g
Beilstein 1310113
Formula Weight 183.25
Percent Purity 97%
Chemical Name or Material Ethyl (E)-3-(1-pyrrolidinyl)crotonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.