missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (ethoxymethylene)cyanoacetate, 98%

Product Code. 10667963 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10667963 1 kg Glass bottle 1kg
10626882 100 g Glass bottle 100g
10636882 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10667963 Supplier Thermo Scientific Acros Supplier No. 118480010
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 94-05-3 | C8H11NO3 | 169.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-05-3
Molecular Formula C8H11NO3
Molecular Weight (g/mol) 169.18
MDL Number MFCD00009136
InChI Key KTMGNAIGXYODKQ-SREVYHEPSA-N
Synonym ethyl ethoxymethylene cyanoacetate, e-ethyl 2-cyano-3-ethoxyacrylate, ethyl 2-cyano-3-ethoxyacrylate, ethyl 3-ethoxy-2-cyanoacrylate, ethyl 2e-2-cyano-3-ethoxyprop-2-enoate, ethyl cyano ethoxymethylene acetate, 2-propenoic acid, 2-cyano-3-ethoxy-, ethyl ester, 2-cyano-3-ethoxy-2-propenoic acid ethyl ester, 2-cyano-3-ethoxyacrylic acid ethyl ester, ethyl 2e-2-cyano-3-ethoxy-2-propenoate
PubChem CID 1715183
SMILES CCO\C=C(\C#N)C(=O)OCC

Specifications

Melting Point 48°C to 52°C
Color White to Yellow
Boiling Point 190°C to 191°C (30.0 mmHg)
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OCH=C(CN)CO2C2H5
Quantity 1 kg
Beilstein 03,47
Fieser 01,38
Solubility Information Solubility in water: <0.01g/L (20°C)
IUPAC Name ethyl (E)-2-cyano-3-ethoxyprop-2-enoate
Formula Weight 169.18
Percent Purity 98%
Physical Form Crystalline Mass or Crystals
Chemical Name or Material Ethyl (ethoxymethylene)cyanoacetate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl (ethoxymethylene)cyanoacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.