missing translation for 'onlineSavingsMsg'
Learn More

Ethyl fluoroacetate, 97%, Thermo Scientific Chemicals

Product Code. 10665042 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10665042 50 g Glass bottle 50g
10439310 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10665042 Supplier Thermo Scientific Acros Supplier No. 172660500
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-72-3
Molecular Formula C4H7FO2
Molecular Weight (g/mol) 106.1
MDL Number MFCD00000450
InChI Key VCYZVXRKYPKDQB-UHFFFAOYSA-N
Synonym ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester
PubChem CID 9988
IUPAC Name ethyl 2-fluoroacetate
SMILES CCOC(=O)CF

Specifications

Color Colorless
Density 1.0900g/mL
Boiling Point 119.2°C (753.0 mmHg)
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula FCH2CO2C2H5
Refractive Index 1.3745 to 1.3765
Quantity 50 g
Beilstein 02, 193
Fieser 05,304
Merck Index 15, 4199
Specific Gravity 1.09
Solubility Information Solubility in water: decomposes.
Formula Weight 106.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl fluoroacetate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl fluoroacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.