missing translation for 'onlineSavingsMsg'
Learn More

Ethyl formate, 98+%, Extra Dry, AcroSeal™

Product Code. 11954891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11954891 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11954891 Supplier Thermo Scientific Acros Supplier No. 429241000
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-94-4 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 109-94-4
Melting Point -80.0°C
Density 0.9170g/mL
Boiling Point 54.0°C
Flash Point −20°C
Assay Percent Range 98% min. (GC)
Packaging AcroSeal™ Glass bottle
Molecular Formula C3H6O2
Linear Formula HCO2C2H5
Refractive Index 1.3580 to 1.3610
MDL Number MFCD00003294
Quantity 100 mL
Merck Index 15,3862
Specific Gravity 0.917
Synonym areginal, formic acid, ethyl ester, ethyl methanoate, formic acid ethyl ester, aethylformiat, ethyl formic ester, ethylformate, ethylformiaat, formic ether, ethylformiat
Solubility Information Solubility in water: 110g/L (18°C). Other solubilities: freely soluble in ethylacetaat, isobutylacetaat,, butylacetaat, aceton, methylisobutylketon and, methylethylketon, miscible with benzene, ether and alcohol
InChI Key WBJINCZRORDGAQ-UHFFFAOYSA-N
SMILES CCOC=O
IUPAC Name ethyl formate
Molecular Weight (g/mol) 74.08
PubChem CID 8025
ChEBI CHEBI:52342
Viscosity 0.4 mPa.s (20°C)
Formula Weight 74.08
Percent Purity 98+%
Grade Extra Dry
Chemical Name or Material Ethyl formate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl formate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.