missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Formate, TRC

Product Code. 30371679
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371679 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371679 Supplier Toronto Research Chemicals Supplier No. TRCE9182002G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Formic acid ethyl ester
CAS 109-94-4
Molecular Formula C3 H6 O2
Quantity 2 g
Synonym Formic Acid Ethyl Ester, Formic acid, ethyl ester, Areginal, Ethyl formate, Ethyl methanoate, Methanoic acid ethyl ester, NSC 406578, NSC 8828
InChI Formula InChI=1S/C3H6O2/c1-2-5-3-4/h3H,2H2,1H3
SMILES CCOC=O
IUPAC Name ethyl formate
Molecular Weight (g/mol) 74.08
Formula Weight 74.0368
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.