missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Gallate, TRC

Product Code. 30137600
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137600 100 g 100g
17958024 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137600 Supplier Toronto Research Chemicals Supplier No. TRCE918320100G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 831-61-8
Molecular Formula C9 H10 O5
Molecular Weight (g/mol) 198.17
Synonym Benzoic acid, 3,4,5-trihydroxy-, ethyl ester, Gallic acid, ethyl ester (6CI,8CI), Phyllemblin (7CI), 3,4,5-Trihydroxybenzoic acid ethyl ester, Ethyl 3,4,5-trihydroxybenzoate, Ethyl gallate, NSC 402626, Nipa No. 48, Nipagallin A, Progallin A
IUPAC Name ethyl 3,4,5-trihydroxybenzoate
SMILES CCOC(=O)c1cc(O)c(O)c(O)c1

Specifications

InChI Formula InChI=1S/C9H10O5/c1-2-14-9(13)5-3-6(10)8(12)7(11)4-5/h3-4,10-12H,2H2,1H3
Formula Weight 198.0528
Quantity 100 g
Chemical Name or Material Ethylgallate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.