missing translation for 'onlineSavingsMsg'
Learn More

Ethyl glyoxylate, ca 50% soln. in toluene

Product Code. 11344076
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344076 100 g 100g
11354076 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344076 Supplier Thermo Scientific Alfa Aesar Supplier No. L19207.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 924-44-7 | C4H6O3 | 102.089 g/mol

Ethyl glyoxylate is widely used as an intermediate in the synthesis of pharmaceuticals (high reactivity of aldehyde function). It is used in the synthesis of a biodegradable polymer, poly(ethyl glyoxylate). It is actively involved in the Friedel-Crafts alkylation reactions with thiophenes to give the corresponding secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl glyoxylate is widely used as an intermediate in the synthesis of pharmaceuticals (high reactivity of aldehyde function). It is used in the synthesis of a biodegradable polymer, poly(ethyl glyoxylate). It is actively involved in the Friedel-Crafts alkylation reactions with thiophenes to give the corresponding secondary alcohols.

Solubility
Immiscible with water.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 924-44-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00044009
InChI Key DBPFRRFGLYGEJI-UHFFFAOYSA-N
Synonym ethyl glyoxalate, ethyl glyoxylate, ethyl oxoacetate, glyoxylic acid ethyl ester, acetic acid, oxo-, ethyl ester, ethyl glyoxalate solution, unii-ixx9mq1j1i, acetic acid, 2-oxo-, ethyl ester, ixx9mq1j1i, 2-oxoacetic acid, ethyl ester
PubChem CID 70211
ChEBI CHEBI:53275
IUPAC Name ethyl 2-oxoacetate
SMILES CCOC(=O)C=O

Specifications

Density 1.003
Boiling Point 110°C
Flash Point 7°C (44°F)
Refractive Index 1.475
Quantity 100 g
UN Number UN1993
Beilstein 1209486
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 102.09
Concentration or Composition (by Analyte or Components) ≈50% soln. in toluene
Chemical Name or Material Ethyl glyoxylate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl glyoxylate, ca 50% solution in toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: toluene, oxo-aceticacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.