missing translation for 'onlineSavingsMsg'
Learn More

Ethyl glyoxylate, ca 50% soln. in toluene

Product Code. 11354076
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11354076

Brand: Thermo Scientific Alfa Aesar L19207.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 924-44-7 | C4H6O3 | 102.089 g/mol

Ethyl glyoxylate is widely used as an intermediate in the synthesis of pharmaceuticals (high reactivity of aldehyde function). It is used in the synthesis of a biodegradable polymer, poly(ethyl glyoxylate). It is actively involved in the Friedel-Crafts alkylation reactions with thiophenes to give the corresponding secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl glyoxylate is widely used as an intermediate in the synthesis of pharmaceuticals (high reactivity of aldehyde function). It is used in the synthesis of a biodegradable polymer, poly(ethyl glyoxylate). It is actively involved in the Friedel-Crafts alkylation reactions with thiophenes to give the corresponding secondary alcohols.

Solubility
Immiscible with water.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 924-44-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00044009
InChI Key DBPFRRFGLYGEJI-UHFFFAOYSA-N
Synonym ethyl glyoxalate, ethyl glyoxylate, ethyl oxoacetate, glyoxylic acid ethyl ester, acetic acid, oxo-, ethyl ester, ethyl glyoxalate solution, unii-ixx9mq1j1i, acetic acid, 2-oxo-, ethyl ester, ixx9mq1j1i, 2-oxoacetic acid, ethyl ester
PubChem CID 70211
ChEBI CHEBI:53275
IUPAC Name ethyl 2-oxoacetate
SMILES CCOC(=O)C=O

Specifications

Density 1.003
Boiling Point 110°C
Flash Point 7°C (44°F)
Refractive Index 1.475
Quantity 500 g
UN Number UN1993
Beilstein 1209486
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 102.09
Concentration or Composition (by Analyte or Components) ≈50% soln. in toluene
Chemical Name or Material Ethyl glyoxylate
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Ethyl glyoxylate, ca 50% solution in toluene
Signalwort
  • Danger
Gefahrenkategorie
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Ergänzende Informationen
  • MIXTURE LIST-Contains : Toluene, Ethyl glyoxylate

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt