missing translation for 'onlineSavingsMsg'
Learn More

Ethyl heptafluorobutyrate, 98%

Product Code. 11452758
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11452758

Brand: Thermo Scientific Alfa Aesar A16742.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 356-27-4 | C6H5F7O2 | 242.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 356-27-4
Molecular Formula C6H5F7O2
Molecular Weight (g/mol) 242.093
MDL Number MFCD00000434
InChI Key JVHJRIQPDBCRRE-UHFFFAOYSA-N
Synonym ethyl heptafluorobutyrate, ethyl perfluorobutyrate, heptafluorobutyric acid ethyl ester, butanoic acid, heptafluoro-, ethyl ester, ethyl heptafluorobutanoate, ethyl perfluorobutanoate, ethyl heptafluoro-n-butyrate, butyric acid, heptafluoro-, ethyl ester, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, ethyl ester, pubchem12539
PubChem CID 9644
IUPAC Name ethyl 2,2,3,3,4,4,4-heptafluorobutanoate
SMILES CCOC(=O)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Boiling Point 95°C to 98°C
Flash Point 14°C (57°F)
Quantity 25 g
UN Number UN3272
Beilstein 1794634
Formula Weight 242.09
Percent Purity 98%
Chemical Name or Material Ethyl heptafluorobutyrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.