missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hydrazinoacetate hydrochloride, 97%

Product Code. 15416518
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15416518

Brand: Thermo Scientific Alfa Aesar H60042.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6945-92-2 | C4H11ClN2O2 | 154.59 g/mol

Ethyl hydrazinoacetate hydrochloride has been used in the preparation of three novel copper(II) complexes as condensation derivatives of 2-pyridinecarboxaldehyde, 2-acetylpyridine and 2-quinolinecarboxaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl hydrazinoacetate hydrochloride has been used in the preparation of three novel copper(II) complexes as condensation derivatives of 2-pyridinecarboxaldehyde, 2-acetylpyridine and 2-quinolinecarboxaldehyde.

Notes
Store in cool dry conditions. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6945-92-2
Molecular Formula C4H11ClN2O2
Molecular Weight (g/mol) 154.59
MDL Number MFCD00012923
InChI Key HZZRIIPYFPIKHR-UHFFFAOYSA-N
Synonym ethyl hydrazinoacetate hydrochloride, ethyl 2-hydrazinylacetate hydrochloride, ethyl hydrazinoacetate hcl, hydrazino-acetic acid ethyl ester hcl, 2-hydrazinyl acetic acid ethyl ester hydrochloride, ethyl 2-hydrazinylacetate hcl, ethyl 2-hydrazinoacetate hydrochloride, acetic acid, hydrazino-, ethyl ester, monohydrochloride, acetic acid, ethyl ester, monohydrochloride, c4h10n2o2.clh
PubChem CID 11593468
IUPAC Name ethyl 2-hydrazinylacetate;hydrochloride
SMILES [H+].[Cl-].CCOC(=O)CNN

Specifications

Melting Point 152°C to 154°C
Linear Formula H2NNHCH2CO2C2H5·HCl
Quantity 25 g
Beilstein 3912112
Formula Weight 154.6
Percent Purity 97%
Chemical Name or Material Ethyl hydrazinoacetate hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.