missing translation for 'onlineSavingsMsg'
Learn More

Ethyl imidazo[2,1-b][1,3]thiazole-6-carboxylate, 97%, Thermo Scientific™

Product Code. 10507303
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10507303 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10507303 Supplier Thermo Scientific Maybridge Supplier No. CC32523DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 64951-04-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H8N2O2S
MDL Number MFCD03419464
Quantity 1 g
Synonym ethyl imidazo 2,1-b thiazole-6-carboxylate, ethyl imidazo 2,1-b 1,3 thiazole-6-carboxylate, imidazo 2,1-b thiazole-6-carboxylic acid ethyl ester, ethylimidazo 2,1-b thiazole-6-carboxylate, ethyl imidazo 2,1-b 1,3-thiazoline-6-carboxylate, imidazo 2,1-, a thiazole-6-carboxylicacid, ethyl ester, imidazo 2,1-b thiazole-6-carboxylicacid, ethyl ester
InChI Key PIURXAHSTOGOSU-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CN2C=CSC2=N1
IUPAC Name ethyl imidazo[2,1-b][1,3]thiazole-6-carboxylate
Molecular Weight (g/mol) 196.224
PubChem CID 7018386
Formula Weight 196.23
Percent Purity 97%
Chemical Name or Material Ethyl imidazo[2, 1-b][1, 3]thiazole-6-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.