missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isobutyrylacetate, 95%

Product Code. 10509043 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10509043 50 g Glass bottle 50g
10413612 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10509043 Supplier Thermo Scientific Acros Supplier No. 383440500
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7152-15-0 | C8H14O3 | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7152-15-0
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.2
MDL Number MFCD00009198
InChI Key XCLDSQRVMMXWMS-UHFFFAOYSA-N
Synonym ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate
PubChem CID 81583
IUPAC Name ethyl 4-methyl-3-oxopentanoate
SMILES CCOC(=O)CC(=O)C(C)C

Specifications

Melting Point -9.0°C
Color Colorless to Yellow
Density 0.9810g/mL
Boiling Point 173.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Mixture of tautomers) (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCOCH2CO2C2H5
Refractive Index 1.4255 to 1.4275
Quantity 50 g
Specific Gravity 0.981
Formula Weight 158.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl isobutyrylacetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl isobutyrylacetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.