missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isodehydroacetate, TRC

Product Code. 30369209
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30369209 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369209 Supplier Toronto Research Chemicals Supplier No. TRCE92135010G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Isodehydroacetate
CAS 3385-34-0
Molecular Formula C10H12O4
Quantity 10 g
Synonym 4,6-Dimethylcoumalic Acid Ethyl Ester, 4-(1-Hydroxyethylidene)-3-methylglutaconic Acid delta-Lactone Ethyl Ester, 5-(Ethoxycarbonyl)-4,6-dimethyl-alpha-pyrone, Ethyl 4,6-Dimethyl-5-coumalate, Ethyl Isodehydracetate, Ethyl Isodehydroacetate, NSC 18911, NSC 602920
InChI Formula InChI=1S/C10H12O4/c1-4-13-10(12)9-6(2)5-8(11)14-7(9)3/h5H,4H2,1-3H3
SMILES CCOC(=O)C1=C(C)OC(=O)C=C1C
IUPAC Name ethyl 2,4-dimethyl-6-oxopyran-3-carboxylate
Molecular Weight (g/mol) 196.2
Formula Weight 196.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.