missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isonipecotate, TRC

Product Code. 30372522
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30372522 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372522 Supplier Toronto Research Chemicals Supplier No. TRCE9214051G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Isonipecotate
CAS 1126-09-6
Molecular Formula C8H15NO2
Quantity 1 g
Synonym 4-Piperidinecarboxylic Acid Ethyl Ester, Isonipecotic Acid Ethyl Ester, 4-(Ethoxycarbonyl)piperidine, 4-Carboethoxypiperidine, Ethyl 4-piperidinecarboxylate, NSC 93792, Piperidin-4-carboxylic Acid Ethyl Ester
InChI Formula InChI=1S/C8H15NO2/c1-2-11-8(10)7-3-5-9-6-4-7/h7,9H,2-6H2,1H3
SMILES CCOC(=O)C1CCNCC1
IUPAC Name ethyl piperidine-4-carboxylate
Molecular Weight (g/mol) 157.21
Formula Weight 157.1103
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.