missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isoquinoline-3-carboxylate, 97%, Thermo Scientific™

Product Code. 10420934
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10420934 1 g Amber Glass Bottle 1g
10596694 5 g Amber Glass Bottle 5g
10759395 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10420934 Supplier Thermo Scientific Maybridge Supplier No. MO08416DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 50458-79-2
Molecular Formula C12H11NO2
Molecular Weight (g/mol) 201.225
MDL Number MFCD10566077
InChI Key IFSCYCNNAIADLI-UHFFFAOYSA-N
Synonym ethyl 3-isoquinolinecarboxylate, ethylisoquinoline-3-carboxylate, ethyl isoquinolin-3-carboxylate, 3-isoquinolinecarboxylic acid, ethyl ester, isoquinoline-3-carboxylic acid ethyl ester, 3-carbethoxyisoquinoline, ksc493m9t, 3-ethoxycarbonyl isoquinoline, 3-isoquinolinecarboxylic acid ethyl ester
PubChem CID 11435602
IUPAC Name ethyl isoquinoline-3-carboxylate
SMILES CCOC(=O)C1=CC2=CC=CC=C2C=N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 201.22
Percent Purity 97%
Chemical Name or Material Ethyl isoquinoline-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.