missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isothiocyanate, 96%

Product Code. 10227602 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10227602 25 g Glass bottle 25g
10025742 5 g Glass bottle 5g
10724014 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10227602 Supplier Thermo Scientific Acros Supplier No. 376860250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 542-85-8 | C3H5NS | 87.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-85-8
Molecular Formula C3H5NS
Molecular Weight (g/mol) 87.14
MDL Number MFCD00004820
InChI Key HBNYJWAFDZLWRS-UHFFFAOYSA-N
Synonym ethyl isothiocyanate, ethane, isothiocyanato, ethyl mustard oil, ethylisothiocyanate, isothiocyanic acid, ethyl ester, unii-3284mj2t8p, ccris 7323, ethylisothio-cyanate, isothiocyanato-ethane
PubChem CID 10966
ChEBI CHEBI:85098
IUPAC Name isothiocyanatoethane
SMILES CCN=C=S

Specifications

Melting Point -6°C
Color Colorless to Yellow
Density 0.9950g/mL
Boiling Point 130°C to 132°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C2H5NCS
Refractive Index 1.5100 to 1.513
Quantity 25 g
Merck Index 15, 3870
Specific Gravity 0.995
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Formula Weight 87.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Ethyl isothiocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.