missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isothiocyanate, 97%

Product Code. 11444067
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444067 25 g 25g
11454067 100 g 100g
11464067 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444067 Supplier Thermo Scientific Alfa Aesar Supplier No. A19492.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 542-85-8 | C3H5NS | 87.14 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Reacts with water. Soluble in chlorinated and aromatic hydrocarbons. Miscible in ethanol and ether.

Notes
Moisture sensitive. Store away from alcohols, amines, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-85-8
Molecular Formula C3H5NS
Molecular Weight (g/mol) 87.14
MDL Number MFCD00004820
InChI Key HBNYJWAFDZLWRS-UHFFFAOYSA-N
Synonym ethyl isothiocyanate, ethane, isothiocyanato, ethyl mustard oil, ethylisothiocyanate, isothiocyanic acid, ethyl ester, unii-3284mj2t8p, ccris 7323, ethylisothio-cyanate, isothiocyanato-ethane
PubChem CID 10966
ChEBI CHEBI:85098
IUPAC Name isothiocyanatoethane
SMILES CCN=C=S

Specifications

Melting Point -6°C
Density 0.995
Boiling Point 130°C to 132°C
Flash Point 32°C (89°F)
Odor Pungent
Refractive Index 1.511
Quantity 25 g
UN Number UN2929
Beilstein 1737705
Sensitivity Moisture sensitive; May form a precipitate
Merck Index 14,3815
Solubility Information Reacts with water. Soluble in chlorinated and aromatic hydrocarbons. Miscible in ethanol and ether.
Formula Weight 87.14
Percent Purity 97%
Chemical Name or Material Ethyl isothiocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.