missing translation for 'onlineSavingsMsg'
Learn More

Ethyl L(-)-lactate 95%, ACROS Organics™

Product Code. 10596782 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10596782 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10596782 Supplier Thermo Scientific Acros Supplier No. 410161000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 687-47-8
Melting Point -26.0°C
Color Colorless to Yellow
Density 1.0340g/mL
Boiling Point 154.0°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C5H10O3
Linear Formula CH3CH(OH)CO2C2H5
Refractive Index 1.4100 to 1.4160
MDL Number MFCD00004518
Quantity 100 g
Beilstein 03, 264
Fieser 17,135
Merck Index 14, 3817
Specific Gravity 1.034
Specific Rotation Condition − 10.00 (20.00°C neat)
Specific Rotation − 10.00
Synonym ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s
InChI Key LZCLXQDLBQLTDK-BYPYZUCNSA-N
SMILES CCOC(=O)C(C)O
IUPAC Name ethyl (2S)-2-hydroxypropanoate
Molecular Weight (g/mol) 118.132
PubChem CID 92831
ChEBI CHEBI:78322
Formula Weight 118.13
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl L(-)-lactate, 95%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl L(-)-lactate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.