missing translation for 'onlineSavingsMsg'
Learn More

Ethyl L-nipecotate, 97%

Product Code. 15463718
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463718 5 g 5g
15473718 250 mg 250mg
15453718 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15463718 Supplier Thermo Scientific Alfa Aesar Supplier No. H57185.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 37675-18-6 | C8H15NO2 | 157.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37675-18-6
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00792499
InChI Key XIWBSOUNZWSFKU-ZETCQYMHSA-N
Synonym s-ethyl piperidine-3-carboxylate, ethyl 3s-piperidine-3-carboxylate, ethyl s-nipecotate, s-+-nipecotic acid ethyl ester, s-ethyl nipecotate, ethyl s-piperidine-3-carboxylate, 3-piperidinecarboxylic acid, ethyl ester, 3s, s-+-ethyl nipecotate, ethyl s-+-nipecotate, s-nipecotic acid ethyl ester
PubChem CID 187784
IUPAC Name ethyl (3S)-piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1

Specifications

Density 1.043
Flash Point 76°C (169°F)
Refractive Index 1.47
Quantity 5 g
Sensitivity Air, light, and moisture sensitive
Optical Rotation +1.5° (Neat)
Formula Weight 157.21
Percent Purity 97%
Chemical Name or Material Ethyl L-nipecotate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.