missing translation for 'onlineSavingsMsg'
Learn More

Ethyl mesitylacetate, 97%

Product Code. 11368075
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11368075

Brand: Thermo Scientific Alfa Aesar L08275.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5460-08-2 | C13H18O2 | 206.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5460-08-2
MDL Number MFCD00051916

Specifications

Density 1.034
Boiling Point 135°C to 140°C (9 mmHg)
Molecular Formula C13H18O2
Refractive Index 1.505
Quantity 2 g
Beilstein 2105791
Synonym ethyl mesitylacetate, ethyl 2-2,4,6-trimethylphenyl acetate, benzeneacetic acid, 2,4,6-trimethyl-, ethyl ester, ethyl 2-mesitylacetate, acmc-20akx5, mesitylacetic acid ethyl ester, ethyl 2,4,6-trimethylphenylacetate, ethyl 2,4,6-trimethylphenyl acetate, 2,4,6-trimethyl-phenyl-acetic acid ethyl ester, benzeneacetic acid,2,4,6-trimethyl-, ethyl ester
InChI Key OFZZPFNYAMIBNU-UHFFFAOYSA-N
SMILES CCOC(=O)CC1=C(C=C(C=C1C)C)C
IUPAC Name ethyl 2-(2,4,6-trimethylphenyl)acetate
Molecular Weight (g/mol) 206.29
PubChem CID 222429
Formula Weight 206.29
Percent Purity 97%
Chemical Name or Material Ethyl mesitylacetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.