missing translation for 'onlineSavingsMsg'
Learn More

Ethyl methacrylate, 99%, stabilized

Product Code. 15720839 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15720839 500 g 500g
10092920 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15720839 Supplier Thermo Scientific Acros Supplier No. 215835000
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 97-63-2 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-63-2
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00009161
InChI Key SUPCQIBBMFXVTL-UHFFFAOYSA-N
Synonym ethyl methacrylate, ethyl 2-methylacrylate, 2-propenoic acid, 2-methyl-, ethyl ester, methacrylic acid ethyl ester, ethyl 2-methyl-2-propenoate, ethyl 2-methacrylate, rhoplex ac-33, methacrylic acid, ethyl ester, rcra waste number u118, 2-methylacrylic acid, ethyl ester
PubChem CID 7343
IUPAC Name ethyl 2-methylprop-2-enoate
SMILES CCOC(=O)C(=C)C

Specifications

Melting Point -75.0°C
Color Colorless
Density 0.9100g/mL
Boiling Point 118.0°C to 119.0°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2C2H5
Refractive Index 1.4130 to 1.4150
Quantity 500 g
Beilstein 02, 423
Specific Gravity 0.91
Solubility Information Solubility in water: 4g/L in water (20°C). Other solubilities: soluble in acetone,slightly soluble in chloroform,miscible in ether and ethanol
Stabilizer 10 to 20ppm MEHQ
Viscosity 0.63 mPa.s (20°C)
Formula Weight 114.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl methacrylate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl methacrylate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl methacrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.