missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Methanesulfonate, TRC

Product Code. 30137854
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137854 100 g 100g
30137264 25 g 25g
18011150 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137854 Supplier Toronto Research Chemicals Supplier No. TRCE917995100G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-50-0
Molecular Formula C3 H8 O3 S
Molecular Weight (g/mol) 124.16
Synonym Methanesulfonic acid, ethyl ester, EMS, Ethyl mesylate, Ethyl methanesulfonate, Ethyl methanesulphonate, NSC 26805, O-Ethyl methylsulfonate
IUPAC Name ethyl methanesulfonate
SMILES CCOS(=O)(=O)C

Specifications

Quantity 100 g
InChI Formula InChI=1S/C3H8O3S/c1-3-6-7(2,4)5/h3H2,1-2H3
Formula Weight 124.0194
Chemical Name or Material Ethyl methanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.