Learn More
Chem-Lab Ethyl methyl ketone ar (MEK) (
Brand:  Chem-Lab CL00.0513.1000
Ethyl methyl ketone ar (MEK) (
Product Code. 13238213
  • ONLINE EXCLUSIVE PRICE
    48.85 €
    42.50 € / 1 L
    Save 6.35 €
    12% Off
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Ethyl methyl ketone a.r. (MEK)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : methyl ethyl ketone