missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Methyl Ketone, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7073020 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Unit Size:
1 L
2.5 L
25 L
5 L
500mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11468253 500 mL Amber glass bottle 500mL
10050160 1 L Amber glass bottle 1 L
10355800 2.5 L Amber glass bottle 2.5 L
10010130 2.5 L Plastic Coated Glass bottle 2.5 L
10487250 5 L Metal can 5 L
10102050 25 L Metal can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11468253 Supplier Fisher Chemical Supplier No. E/1450/08
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C4H8O, CAS Number-78-93-3, methyl ethyl ketone, methylethylketone, methyl acetone, 2-butanone, methylethyl ketone, meetco, butanone, 3-butanone, ethyl methyl ketone, ethylmethylketon, 2.5L, 80 deg.C, CHEBI:28398, Colorless, 72.11g/mol, ZWEHNKRNPOVVGH-UHFFFAOYSA-N, butan-2-one, -87 deg.C, 11648

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -87°C
Color Colorless
Boiling Point 80°C
Packaging Amber glass bottle
Quantity 500 mL
Formula Weight 72.11g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • 2-Butanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P240-Ground/bond container and receiving equipment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.