missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 97%, tech.

Product Code. 10780661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10780661 5 g Glass bottle 5g
10571221 25 g Glass bottle 25g
10398243 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10780661 Supplier Thermo Scientific Acros Supplier No. 162300050
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52763-21-0 | C15H20ClNO3 | 297.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52763-21-0
Molecular Formula C15H20ClNO3
Molecular Weight (g/mol) 297.78
MDL Number MFCD00012792
InChI Key UQOMEAWPKSISII-UHFFFAOYNA-N
Synonym ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, ethyl n-benzyl-3-oxo-4-piperidine-carboxylate hydrochloride, ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 1-benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hcl, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hydrochloride, 1-benzyl-3-oxopiperidine-4-carboxylic acid ethyl ester hydrochloride, ethyl n-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, zlchem 420, pubchem19671
PubChem CID 2723880
IUPAC Name ethyl 1-benzyl-3-oxopiperidine-4-carboxylate;hydrochloride
SMILES [H+].[Cl-].CCOC(=O)C1CCN(CC2=CC=CC=C2)CC1=O

Specifications

Melting Point 160°C to 165°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 297.77
Percent Purity 97%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl N-benzyl-3-oxo-4-piperidine-carboxylate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.