missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N-hydroxyacetimidate, 97%

Product Code. 11404473
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404473 100 g 100g
11484463 5 g 5g
11494463 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404473 Supplier Thermo Scientific Alfa Aesar Supplier No. A11006.22
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10576-12-2 | C4H9NO2 | 103.12 g/mol

Ethyl N-hydroxyacetimidate was used in the synthesis of O-acyl-and O-nitrophenylhydroxylamines, hydroxamic acid ethoxycarbonylhydrazides and ethyl O-(2,4-dinitrophenyl)acetohydroxamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl N-hydroxyacetimidate was used in the synthesis of O-acyl-and O-nitrophenylhydroxylamines, hydroxamic acid ethoxycarbonylhydrazides and ethyl O-(2,4-dinitrophenyl)acetohydroxamate.

Solubility
Decompose in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10576-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00002114,MFCD00002114
InChI Key QWKAVVNRCKPKNM-SNAWJCMRSA-N
Synonym ethyl n-hydroxyacetimidate, n-hydroxyethanimidic acid, ethyl ester, ethyl acetohydroximate, ethyl n-hydroxyethanimidate, e-ethyl n-hydroxyethanimidate, e-ethyl n-hydroxyacetimidate, ethyl 1e-n-hydroxyethanimidate, snx`lfelmmpqh, ethyl acethydroxamate, 1-ethoxyethanone oxime
PubChem CID 6386647
IUPAC Name ethyl (1Z)-N-hydroxyethanimidate
SMILES CCO\C(C)=N\O

Specifications

Melting Point 23°C to 25°C
Boiling Point 55°C to 58°C (6 mmHg)
Flash Point 77°C (171°F)
Refractive Index 1.434
Quantity 100 g
Beilstein 1700260
Sensitivity Moisture sensitive
Solubility Information Decompose in water.
Formula Weight 103.12
Percent Purity 97%
Chemical Name or Material Ethyl N-hydroxyacetimidate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.