missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N-piperazinecarboxylate, 99%

Product Code. 10205110 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
This item is not returnable. View return policy

Product Code. 10205110

Brand: Thermo Scientific Acros 118870250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-43-4 | C7H14N2O2 | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-43-4
Molecular Formula C7H14N2O2
Molecular Weight (g/mol) 158.2
MDL Number MFCD00005964
InChI Key LNOQURRKNJKKBU-UHFFFAOYSA-N
Synonym ethyl n-piperazinecarboxylate, ethyl 1-piperazinecarboxylate, n-carbethoxypiperazine, 1-carbethoxypiperazine, 1-carboethoxypiperazine, 1-piperazinecarboxylic acid, ethyl ester, n-ethoxycarbonyl piperazine, ethylcarbonyl piperazine, 1-ethoxycarbonyl piperazine, ethyl 1-piperazinocarboxylate
PubChem CID 8431
IUPAC Name ethyl piperazine-1-carboxylate
SMILES CCOC(=O)N1CCNCC1

Specifications

Color Yellow
Density 1.0800g/mL
Boiling Point 273°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4760 to 1.478
Quantity 25 g
Beilstein 23, II, 9
Specific Gravity 1.08
Solubility Information Solubility in water: freely soluble
Formula Weight 158.2
Percent Purity 99%
Physical Form Oily Liquid
Chemical Name or Material Ethyl N-piperazinecarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl N-piperazinecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.