missing translation for 'onlineSavingsMsg'
Learn More

Ethyl nipecotate, 97%, Thermo Scientific™

Product Code. 11481167
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481167 100 g 100g
11471167 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481167 Supplier Thermo Scientific Alfa Aesar Supplier No. B24155.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5006-62-2
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00005991
InChI Key XIWBSOUNZWSFKU-UHFFFAOYSA-N
Synonym ethyl nipecotate, ethyl 3-piperidinecarboxylate, nipecotic acid ethyl ester, 3-piperidinecarboxylic acid ethyl ester, ethylpiperidine-3-carboxylate, 3-ethoxycarbonyl piperidine, 3-piperidinecarboxylic acid, ethyl ester, piperidine-3-carboxylic acid ethyl ester, ethylnipecotate, a-ethyl nipecotate
PubChem CID 98969
IUPAC Name ethyl piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1

Specifications

Density 1.012
Boiling Point 102°C to 104°C (7 mmHg)
Flash Point 90°C (195°F)
Refractive Index 1.46
Quantity 100 g
Sensitivity Air Sensitive
Formula Weight 157.21
Percent Purity 97%
Chemical Name or Material Ethyl nipecotate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.