missing translation for 'onlineSavingsMsg'
Learn More

Ethyl oleate, 98%, mixture of homologeous fatty acid esters

Product Code. 10578322 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10578322 250 g Glass bottle 250g
10627191 1 kg Glass bottle 1kg
11964961 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10578322 Supplier Thermo Scientific Acros Supplier No. 410292500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 111-62-6 | C20H38O2 | 310.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 111-62-6
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.51
InChI Key LVGKNOAMLMIIKO-QXMHVHEDSA-N
Synonym ethyl oleate, ethyl cis-9-octadecenoate, oleic acid, ethyl ester, oleic acid ethyl ester, 9-octadecenoic acid z-, ethyl ester, ethyl oleate nf, ethyl z-octadec-9-enoate, ethyl oleate natural, fema no. 2450
PubChem CID 5363269
ChEBI CHEBI:84940
IUPAC Name ethyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC

Spécification

Acidity 0.5mg KOH/g max.
Density 0.8600g/mL
Boiling Point 190.0°C to 210.0°C (7.0 mmHg)
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 60% min. (oleic acid) (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7CHCH(CH2)7COOC2H5
Refractive Index 1.4490 to 1.4530
Quantity 250 g
Saponification Value 177 to 188mg KOH/g
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 310.51
Percent Purity 98%
Physical Form Oily Liquid
Chemical Name or Material Ethyl oleate
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.