missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Oleate, Pure, for Saponification, Fisher Chemical

Product Code. 10786091 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10786091 500 mL Amber glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10786091 Supplier Fisher Chemical Supplier No. E/1460/08
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tekniske data

CAS 111-62-6
Melting Point 10°C
Color Colorless to Yellow
Boiling Point 210°C
Packaging Amber glass bottle
Molecular Formula C20H38O2
MDL Number 9579
Quantity 500 mL
Synonym ethyl oleate, ethyl cis-9-octadecenoate, oleic acid, ethyl ester, oleic acid ethyl ester, 9-octadecenoic acid z-, ethyl ester, ethyl oleate nf, ethyl z-octadec-9-enoate, ethyl oleate natural, fema no. 2450
InChI Key LVGKNOAMLMIIKO-QXMHVHEDSA-N
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC
IUPAC Name ethyl (Z)-octadec-9-enoate
Molecular Weight (g/mol) 310.522
PubChem CID 5363269
ChEBI CHEBI:84940
Formula Weight 310.52g/mol
Physical Form Liquid
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.