missing translation for 'onlineSavingsMsg'
Learn More

Ethyl oxalyl chloride, 98%

Product Code. 11457616
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457616 500 g 500g
11437616 25 g 25g
11447616 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457616 Supplier Thermo Scientific Alfa Aesar Supplier No. A14653.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4755-77-5 | C4H5ClO3 | 136.531 g/mol

Ethyl oxalyl chloride is used in the synthesis of 2-oxo-3-alkenoic esters and alpha-keto ester in good yield. It acts as a reactant in the preparation of oxyoxalamide derivatives as an epoxide hydrolase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl oxalyl chloride is used in the synthesis of 2-oxo-3-alkenoic esters and alpha-keto ester in good yield. It acts as a reactant in the preparation of oxyoxalamide derivatives as an epoxide hydrolase inhibitor.

Solubility
Slightly miscible with water.

Notes
Moisture sensitive. Incompatible with bases, oxidizing agents and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4755-77-5
Molecular Formula C4H5ClO3
Molecular Weight (g/mol) 136.531
MDL Number MFCD00000706
InChI Key OWZFULPEVHKEKS-UHFFFAOYSA-N
Synonym ethyl chlorooxoacetate, ethyl oxalyl chloride, ethyl chloroglyoxylate, ethoxalyl chloride, ethyl oxalyl monochloride, acetic acid, chlorooxo-, ethyl ester, oxalic acid monoethyl ester chloride, monoethyl oxaloyl chloride, oxalyl chloride, ethyl ester, ethyl chlorocarbonyl formate
PubChem CID 20884
IUPAC Name ethyl 2-chloro-2-oxoacetate
SMILES CCOC(=O)C(=O)Cl

Specifications

Density 1.226
Boiling Point 134°C to 135°C
Flash Point 41°C (105°F)
Linear Formula ClCOCO2CH2CH3
Refractive Index 1.416
Quantity 500 g
UN Number UN2920
Beilstein 506725
Sensitivity Moisture sensitive
Solubility Information Slightly miscible with water.
Formula Weight 136.53
Percent Purity 98%
Chemical Name or Material Ethyl oxalyl chloride
Show More Show Less

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.