missing translation for 'onlineSavingsMsg'
Learn More

Ethyl perfluorooctanoate, 97%, Thermo Scientific™

Product Code. 11410197
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410197 1 g 1g
11420197 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410197 Supplier Thermo Scientific Alfa Aesar Supplier No. B21639.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3108-24-5
Molecular Formula C10H5F15O2
Molecular Weight (g/mol) 442.124
MDL Number MFCD00018063
InChI Key JTOFFHFAQBLPTM-UHFFFAOYSA-N
PubChem CID 76555
IUPAC Name ethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
SMILES CCOC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.626
Boiling Point 167°C to 168°C
Flash Point None
Linear Formula CF3(CF2)6CO2CH2CH3
Refractive Index 1.311
Quantity 1 g
Beilstein 1810008
Synonym ethyl perfluorooctanoate, ethyl perfluorocaprylate, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, ethyl ester, octanoic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, ethyl ester, acmc-20akwv, ethyl perfluorooctanoat, ethyl perfluorooctanonate, octanoic acid, pentadecafluoro-, ethyl ester, pentadecafluorooctanoic acid ethyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-octanoic acid ethyl ester
Formula Weight 442.12
Percent Purity 97%
Chemical Name or Material Ethyl perfluorooctanoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.