missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Perfluorooctanoate, TRC

Product Code. 30041276
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041276 100 mg 100mg
30040384 50 mg 50mg
30040777 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041276 Supplier Toronto Research Chemicals Supplier No. TRCE192630100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3108-24-5
Molecular Formula C10H5F15O2
Molecular Weight (g/mol) 442.13
Synonym Perfluorooctanoic Acid-Ethyl Ester, Ethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluorooctanoate, Ethyl Pentadecafluorooctanoate, Ethyl Ester Pentadecafluorooctanoic Acid;
IUPAC Name ethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
SMILES CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H5F15O2/c1-2-27-3(26)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)9(21,22)10(23,24)25/h2H2,1H3
Formula Weight 442.005
Chemical Name or Material Ethyl Perfluorooctanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.