missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Phenylacetate, TRC

Product Code. 30380618
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380618 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380618 Supplier Toronto Research Chemicals Supplier No. TRCE92581010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylacetic Acid Ethylester
CAS 101-97-3
Molecular Formula C10 H12 O2
Quantity 10 g
Synonym Ethyl 2-phenyl Acetate, Ethyl 2-phenylacetate, Benzeneacetic acid, ethyl ester, Acetic acid, phenyl-, ethyl ester (6CI,8CI), 2-Phenylacetic acid ethyl ester, Ethyl 2-phenylacetate, Ethyl 2-phenylethanoate, Ethyl benzeneacetate, Ethyl phenacetate, Ethyl phenylacetate, Ethyl α-phenylacetate, Ethyl α-toluate, NSC 406259, NSC 8894, Phenylacetic acid ethyl ester, α-Toluic acid ethyl ester
InChI Formula InChI=1S/C10H12O2/c1-2-12-10(11)8-9-6-4-3-5-7-9/h3-7H,2,8H2,1H3
SMILES CCOC(=O)Cc1ccccc1
IUPAC Name ethyl 2-phenylacetate
Molecular Weight (g/mol) 164.2
Formula Weight 164.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.