missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenylcyanoacetate, 97%

Product Code. 10142480 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10142480

Brand: Thermo Scientific Acros 118830050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4553-07-5 | C11H11NO2 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4553-07-5
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00001866
InChI Key SXIRJEDGTAKGKU-UHFFFAOYNA-N
SMILES CCOC(=O)C(C#N)C1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 275°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5CH(CN)CO2C2H5
Refractive Index 1.5033 to 1.5053
Quantity 5 g
Beilstein 09, 854
Specific Gravity 1.09
Synonym s-dibenzyl 2-aminopentanedioate hydrochloride, h-glu obzl-obzl.hcl, l-glutamic acid dibenzyl ester hydrochloride, l-glutamic acid, bis phenylmethyl ester, hydrochloride, s-dibenzyl 2-aminopentanedioate hcl, dibenzyl l-glutaminate hydrochloride, glutamic acid, dibenzyl ester, hydrochloride, l, 1,5-dibenzyl 2s-2-aminopentanedioate hydrochloride, h-glu obzl-obzl.hci, h-glu obzl-obzl hcl
IUPAC Name dibenzyl (2S)-2-aminopentanedioate;hydrochloride
PubChem CID 199613
Formula Weight 189.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl phenylcyanoacetate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl phenylcyanoacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.