missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenylcyanoacetate, 98%

Product Code. 11376716
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11376716 25 g 25g
11366716 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376716 Supplier Thermo Scientific Alfa Aesar Supplier No. L03929.14
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4553-07-5 | C11H11NO2 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4553-07-5
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00001866
InChI Key SXIRJEDGTAKGKU-UHFFFAOYNA-N
Synonym ethyl phenylcyanoacetate, phenylcyanoacetic acid ethyl ester, ethyl cyanophenylacetate, ethyl cyano phenyl acetate, ethyl alpha-cyanophenylacetate, ethyl alpha-cyano-alpha-phenylacetate, acetic acid, cyanophenyl-, ethyl ester, alpha-cyanobenzeneacetic acid, ethyl ester, benzeneacetic acid, alpha-cyano-, ethyl ester, benzeneacetic acid, .alpha.-cyano-, ethyl ester
PubChem CID 95298
ChEBI CHEBI:51926
IUPAC Name ethyl 2-cyano-2-phenylacetate
SMILES CCOC(=O)C(C#N)C1=CC=CC=C1

Specifications

Density 1.09
Boiling Point 274°C to 275°C
Flash Point >110°C (230°F)
Refractive Index 1.505
Quantity 25 g
Beilstein 611232
Formula Weight 189.21
Percent Purity 98%
Chemical Name or Material Ethyl phenylcyanoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.