missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenylpropiolate, 98%

Product Code. 10724421 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10724421 5 g Glass bottle 5g
10580751 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10724421 Supplier Thermo Scientific Acros Supplier No. 118850050
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2216-94-6 | C11H10O2 | 174.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2216-94-6
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.2
MDL Number MFCD00009185
InChI Key ACJOYTKWHPEIHW-UHFFFAOYSA-N
Synonym ethyl phenylpropiolate, ethylphenylpropiolate, ethyl 3-phenylpropiolate, ethyl phenylpropriolate, 2-propynoic acid, 3-phenyl-, ethyl ester, ethyl 3-phenylpropynoate, ethyl phenylpropynoate, ethyl phenylacetylenecarboxylate, phenylpropiolic acid ethyl ester, ethyl 3-phenyl-2-propynoate
PubChem CID 91516
IUPAC Name ethyl 3-phenylprop-2-ynoate
SMILES CCOC(=O)C#CC1=CC=CC=C1

Specifications

Color Yellow
Density 1.0500g/mL
Boiling Point 260°C to 270°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5C∽CCO2C2H5
Refractive Index 1.5510 to 1.553
Quantity 5 g
Beilstein 09, 634
Specific Gravity 1.05
Formula Weight 174.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl phenylpropiolate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.