missing translation for 'onlineSavingsMsg'
Learn More

Ethyl picolinate, 99%

Product Code. 11448930
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
Unit Size:
100g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448930 100 g 100g
11438930 25 g 25g
11411728 50 g 50g
11421728 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11448930 Supplier Thermo Scientific Alfa Aesar Supplier No. A13973.22
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2524-52-9 | C8H9NO2 | 151.17 g/mol

Ethyl picolinate is used in the preparation of 2-Aminodihydro[1,3]thiazines as BACE 2 inhibitors which is used in the treatment of diabetes. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl picolinate is used in the preparation of 2-Aminodihydro[1,3]thiazines as BACE 2 inhibitors which is used in the treatment of diabetes. It is also used as pharmaceutical intermediate.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2524-52-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00006292
InChI Key FQYYIPZPELSLDK-UHFFFAOYSA-N
Synonym ethyl picolinate, ethyl 2-picolinate, ethyl 2-pyridinecarboxylate, picolinic acid, ethyl ester, 2-picolinic acid ethyl ester, 2-pyridinecarboxylic acid, ethyl ester, 2-ethoxycarbonyl pyridine, picolinic acid ethyl ester, 2-pyridinecarboxylic acid ethyl ester, pyridine-2-carboxylic acid ethyl ester
PubChem CID 17307
IUPAC Name ethyl pyridine-2-carboxylate
SMILES CCOC(=O)C1=CC=CC=N1

Specifications

Melting Point 1°C to 3°C
Density 1.12
Boiling Point 239°C to 241°C
Flash Point 107°C (224°F)
Odor Characteristic
Refractive Index 1.5105
Quantity 100 g
Beilstein 122485
Solubility Information Fully miscible in water.
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material Ethyl picolinate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.