missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pipecolinate, 98+%

Product Code. 11398485
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11398485 1 g 1g
11308495 5 g 5g
11318495 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11398485

Brand: Thermo Scientific Alfa Aesar L10532.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15862-72-3 | C8H16NO2 | 158.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15862-72-3
Molecular Formula C8H16NO2
Molecular Weight (g/mol) 158.22
MDL Number MFCD00005980
InChI Key SZIKRGHFZTYTIT-ZETCQYMHSA-O
Synonym ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720
PubChem CID 27517
IUPAC Name ethyl piperidine-2-carboxylate
SMILES CCOC(=O)[C@@H]1CCCC[NH2+]1

Specifications

Density 1.006
Boiling Point 216°C to 217°C
Flash Point 46°C (114°F)
Refractive Index 1.456
Quantity 1 g
UN Number UN1993
Beilstein 81694
Formula Weight 157.21
Percent Purity ≥98%
Chemical Name or Material Ethyl pipecolinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.