missing translation for 'onlineSavingsMsg'
Learn More

Ethyl potassium oxalate, 97%

Product Code. 15447517
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447517 5 g 5g
15457517 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447517 Supplier Thermo Scientific Alfa Aesar Supplier No. H35403.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1906-57-6 | C4H5KO4 | 156.178 g/mol

Ethyl potassium oxalate is used to prepare oxalic acid ethyl ester 2-oxo-3-phenyl-propyl ester by reacting with 1-bromo-3-phenyl-acetone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl potassium oxalate is used to prepare oxalic acid ethyl ester 2-oxo-3-phenyl-propyl ester by reacting with 1-bromo-3-phenyl-acetone.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1906-57-6
Molecular Formula C4H5KO4
Molecular Weight (g/mol) 156.178
MDL Number MFCD00045861
InChI Key RLPQQBNSTHRHEK-UHFFFAOYSA-M
Synonym ethyl potassium oxalate, potassium ethyl oxalate, ethanedioic acid, monoethyl ester, potassium salt, potassium 2-ethoxy-2-oxoacetate, oxalic acid 1-ethyl 2-potassium salt, ethanedioic acid, 1-ethyl ester, potassium salt 1:1, potassium ethoxy oxo acetate, ambkkkkk567, oxalic acid, monoethyl ester, potassium salt, potassium monoethyl oxalate
PubChem CID 23678856
IUPAC Name potassium;2-ethoxy-2-oxoacetate
SMILES CCOC(=O)C(=O)[O-].[K+]

Specifications

Melting Point 220°C to 223°C
Quantity 5 g
UN Number UN2811
Formula Weight 156.18
Percent Purity 97%
Chemical Name or Material Ethyl potassium oxalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.