missing translation for 'onlineSavingsMsg'
Learn More

Ethyl propiolate, 99%

Product Code. 10667262 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10667262

Brand: Thermo Scientific Acros 118900050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-47-2 | C5H6O2 | 98.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-47-2
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00009184
InChI Key FMVJYQGSRWVMQV-UHFFFAOYSA-N
Synonym ethyl propiolate, ethyl acetylenecarboxylate, 2-propynoic acid, ethyl ester, propiolic acid ethyl ester, ethyl propynoate, ethoxycarbonyl acetylene, ethyl 2-propynoate, propiolic acid, ethyl ester, unii-w235g5u52s, propynoic acid ethyl ester
PubChem CID 12182
ChEBI CHEBI:51740
IUPAC Name ethyl prop-2-ynoate
SMILES CCOC(=O)C#C

Specifications

Color Colorless to Yellow
Density 0.9680g/mL
Boiling Point 120°C
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HC∽CCO2C2H5
Refractive Index 1.4110 to 1.413
Quantity 5 g
Beilstein 02, 477
Merck Index 15, 3898
Specific Gravity 0.968
Solubility Information Solubility in water: miscible with water. Other solubilities: soluble in most organic solvents
Formula Weight 98.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl propiolate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.