missing translation for 'onlineSavingsMsg'
Learn More

Ethyl propiolate, 99%

Product Code. 11427357
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11427357

Brand: Thermo Scientific Alfa Aesar A15068.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-47-2 | C5H6O2 | 98.10 g/mol

Ethyl propiolate is used as a precursor in the preparation of substituted anthraquinones and pyrazolo[1,5-a]pyridine, which exhibits anti-inflammatory activity. Further, it is employed in the synthesis of benzene-1,2,3,5-tetracarboxylates promoted by triphenylphosphine. Its halogenated derivatives are used as a substrate for direct, asymmetric alkynylation of cyclic beta-ketoesters using chiral phase-transfer catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl propiolate is used as a precursor in the preparation of substituted anthraquinones and pyrazolo[1,5-a]pyridine, which exhibits anti-inflammatory activity. Further, it is employed in the synthesis of benzene-1,2,3,5-tetracarboxylates promoted by triphenylphosphine. Its halogenated derivatives are used as a substrate for direct, asymmetric alkynylation of cyclic beta-ketoesters using chiral phase-transfer catalysts.

Solubility
Miscible with alcohol.

Notes
Incompatible with oxidizing agents, bases and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-47-2
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00009184
InChI Key FMVJYQGSRWVMQV-UHFFFAOYSA-N
Synonym ethyl propiolate, ethyl acetylenecarboxylate, 2-propynoic acid, ethyl ester, propiolic acid ethyl ester, ethyl propynoate, ethoxycarbonyl acetylene, ethyl 2-propynoate, propiolic acid, ethyl ester, unii-w235g5u52s, propynoic acid ethyl ester
PubChem CID 12182
ChEBI CHEBI:51740
IUPAC Name ethyl prop-2-ynoate
SMILES CCOC(=O)C#C

Specifications

Melting Point 9°C
Density 0.963
Boiling Point 118°C to 120°C
Flash Point 23°C (73°F)
Odor Pungent
Linear Formula HC≡CCO2CH2CH3
Refractive Index 1.412
Quantity 5 g
UN Number UN3272
Beilstein 878250
Solubility Information Miscible with alcohol.
Formula Weight 98.1
Percent Purity 99%
Chemical Name or Material Ethyl propiolate
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Ethyl propiolate
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
  • MIXTURE LIST-Contains : Ethyl propiolate

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt