missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pyrrole-2-carboxylate, 98+%

Product Code. 11339145
Click to view available options
Quantity:
5 g
25 g
missing translation for 'unitSize'
25g
5g
This item is not returnable. View return policy

Product Code. 11339145

missing translation for 'mfr': Thermo Scientific Alfa Aesar L16382.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2199-43-1 | C7H9NO2 | 139.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2199-43-1
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00817049
InChI Key PAEYAKGINDQUCT-UHFFFAOYSA-N
Synonym ethyl pyrrole-2-carboxylate, ethyl 2-pyrrolecarboxylate, ethyl pyrrole-2-carbonate, ethylpyrrole-2-carboxylate, 1h-pyrrole-2-carboxylic acid, ethyl ester, 2-ethoxycarbonyl-1h-pyrrole, pyrrole-2-carboxylic acid, ethyl ester, 1h-pyrrole-2-carboxylic acid ethyl ester, pyrrole-2-carboxylic acid ethyl ester, ethyl1h-pyrrole-2-carboxylate
PubChem CID 255670
IUPAC Name ethyl 1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=CC=CN1

Specifications

Melting Point 37°C to 39°C
Boiling Point 70°C (0.8 mmHg)
Quantity 5 g
Beilstein 2645
Formula Weight 139.16
Percent Purity ≥98%
Chemical Name or Material Ethyl pyrrole-2-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.