missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pyruvate, 98%

Product Code. 11457307
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
missing translation for 'unitSize':
250g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity missing translation for 'unitSize'
11457307 250 g 250g
11447307 50 g 50g
11418930 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11457307 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A15045.30
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 617-35-6 | C5H8O3 | 116.116 g/mol

Ethyl pyruvate is used as a flavoring agent in food and anti-inflammatory agent for the treatment of critical inflammatory conditions. It is used in the treatment of critical illnesses such as severe sepsis, acute respiratory distress syndrome, burn injury, acute pancreatitis and stroke. It plays an important role in cardiac function after coronary ischemia and reperfusion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water, ethanol and ether.

Notes
Store in a cool place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 617-35-6
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00009123
InChI Key XXRCUYVCPSWGCC-UHFFFAOYSA-N
Synonym ethyl pyruvate, ethyl 2-oxopropionate, pyruvic acid, ethyl ester, propanoic acid, 2-oxo-, ethyl ester, pyruvic acid ethyl ester, ethyl pyroracemate, ethyl acetylformate, 2-oxo-propionic acid ethyl ester, ethyl pyruvate natural, ethyl alpha-ketopropionate
PubChem CID 12041
IUPAC Name ethyl 2-oxopropanoate
SMILES CCOC(=O)C(=O)C

Spezifikation

Melting Point -58°C
Density 1.05
Boiling Point 143°C to 145°C
Flash Point 45°C (113°F)
Assay Percent Range 98%
Odor Pleasant
Refractive Index 1.405
Quantity 250 g
UN Number UN3272
Beilstein 1071466
Merck Index 14,8021
Solubility Information Miscible with water,ethanol and ether.
Formula Weight 116.12
Percent Purity 98%
Chemical Name or Material Ethyl pyruvate
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Ethyl pyruvate
Signalwort
  • Warning
Gefahrenkategorie
  • Flammable liquid Category 3
Gefahrenhinweis
  • H226-Flammable liquid and vapour.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Ethyl pyruvate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.