missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-nipecotate L-tartrate, 98%, Thermo Scientific Chemicals

Product Code. 15492668
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15492668 25 g 25g
15472668 1 g 1g
15482668 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492668 Supplier Thermo Scientific Alfa Aesar Supplier No. H56226.14
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167392-57-6
Molecular Formula C12H21NO8
Molecular Weight (g/mol) 307.299
MDL Number MFCD00799538
InChI Key HHPGQKZOPPDLNH-RATVZYDNSA-N
Synonym s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester
PubChem CID 53249980
IUPAC Name (3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 157°C to 159°C
Quantity 25 g
Optical Rotation +10° (c=5 in Water)
Formula Weight 307.3
Percent Purity 98%
Chemical Name or Material Ethyl (R)-nipecotate L-tartrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.