missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%, Thermo Scientific Chemicals

Product Code. 15497346
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497346 5 g 5g
15487346 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497346 Supplier Thermo Scientific Alfa Aesar Supplier No. H62378.06
 Restricted Item
Estimated Shipment: 18-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 144978-12-1 | C12H19NO5 | 257.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 144978-12-1
Molecular Formula C12H19NO5
Molecular Weight (g/mol) 257.29
MDL Number MFCD07778090
InChI Key YWWWGFSJHCFVOW-UHFFFAOYNA-N
Synonym boc-pyr-oet, s-ethyl-n-boc-pyroglutamate, n-boc-l-pyroglutamic acid ethyl ester, 1-boc-l-pyroglutamic acid ethyl ester, boc-l-pyroglutamic acid ethyl ester, s-1-tert-butyl 2-ethyl 5-oxopyrrolidine-1,2-dicarboxylate, ethyl n-boc-l-pyroglutamate, 1-tert-butyl 2-ethyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, s-n-alpha-t-butyloxycarbonyl-pyroglutamic acid ethyl ester, 1-tert-butyl 2-ethyl s-5-oxopyrrolidine-1,2-dicarboxylate
PubChem CID 5461186
SMILES CCOC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C

Tekniske data

Quantity 5 g
IUPAC Name 1-O-tert-butyl 2-O-ethyl (2S)-5-oxopyrrolidine-1,2-dicarboxylate
Formula Weight 257.28
Percent Purity 98%
Chemical Name or Material Ethyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.