missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-2-oxiranylacetate, 97%, Thermo Scientific Chemicals

Product Code. 15473807
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473807 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15473807 Supplier Thermo Scientific Alfa Aesar Supplier No. H27891.ME
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mainly employed inorganic synthesis. They are starting materials for the condensation reactions. Homoallylic alcohol is readily available from the regiospecific ring opening of commercially available ethyl (S)-oxiranyl acetate with
TRUSTED_SUSTAINABILITY

Specifications

CAS 112083-63-3
Molecular Formula C6H10O3
MDL Number MFCD00672889
Quantity 500 mg
Sensitivity Air sensitive
Synonym ethyl s-2-oxiranylacetate, s-ethyl 2-oxiran-2-yl acetate, ethyl s-3,4-epoxybutyrate, s-oxiraneacetic acid ethyl ester, oxiraneacetic acid, ethyl ester, 2s, 2-oxiraneacetic acid,ethyl ester, 2s, ethyl 2-2s-oxiran-2-yl acetate, ethyl s-oxirane-2-acetate, ethyl 2s-oxiran-2-ylacetate, ethyl s-3,4-epoxybutanoate
Solubility Information Decomposes in water.
InChI Key WHUSTVAXKRFVPD-UHFFFAOYNA-N
SMILES CCOC(=O)CC1CO1
IUPAC Name ethyl 2-(oxiran-2-yl)acetate
Molecular Weight (g/mol) 130.14
PubChem CID 10920539
Formula Weight 130.14
Percent Purity 97%
Chemical Name or Material Ethyl (S)-2-oxiranylacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.