missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate, 99%

Product Code. 10266670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10266670 10 g Glass bottle 10g
10592631 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10266670 Supplier Thermo Scientific Acros Supplier No. 233320100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7149-65-7 | C7H11NO3 | 157.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7149-65-7
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.16
MDL Number MFCD00064497
InChI Key QYJOOVQLTTVTJY-YFKPBYRVSA-N
Synonym l-pyroglutamic acid ethyl ester, ethyl s-+-2-pyrrolidone-5-carboxylate, s-ethyl 5-oxopyrrolidine-2-carboxylate, ethyl 2s-5-oxopyrrolidine-2-carboxylate, ethyl l-pyroglutamate, ethyl pidolate, h-pyr-oet, ethyl 5-oxo-l-prolinate, ethyl pyroglutamate, unii-1ps59es316
PubChem CID 2724446
IUPAC Name ethyl (2S)-5-oxopyrrolidine-2-carboxylate
SMILES CCOC(=O)C1CCC(=O)N1

Specifications

Melting Point 47°C to 51°C
Color White to Cream
Boiling Point 176°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Specific Rotation Condition - 3.50 (20.00°C c=5,water)
Specific Rotation -3.5
Formula Weight 157.16
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.