missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate, 98%

Product Code. 11309405
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309405 5 g 5g
11399395 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309405 Supplier Thermo Scientific Alfa Aesar Supplier No. L18554.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86728-85-0 | C6H11ClO3 | 166.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86728-85-0
Molecular Formula C6H11ClO3
Molecular Weight (g/mol) 166.60
MDL Number MFCD00211241
InChI Key ZAJNMXDBJKCCAT-YFKPBYRVSA-N
Synonym ethyl s-4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutanoate, ethyl 3s-4-chloro-3-hydroxybutanoate, ethyl s-4-chloro-3-hydroxybutanoate, s-4-chloro-3-hydroxybutyrate, s-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-n-butyric acid ethyl ester, s-ethyl 4-chloro-3-hydroxybutanoate
PubChem CID 7019277
IUPAC Name ethyl (3S)-4-chloro-3-hydroxybutanoate
SMILES CCOC(=O)C[C@H](O)CCl

Specifications

Density 1.19
Boiling Point 93°C to 95°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.453
Quantity 5 g
Optical Rotation −14° (Neat)
Formula Weight 166.61
Percent Purity 98%
Assay ee 98%
Chemical Name or Material Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.